Z655236
Hanna conductivity standards
conductivity 12880 μS/cm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41100000
Recommended Products
manufacturer/tradename
Hanna HI 70030C
conductivity measuring range
12880 μS/cm
Looking for similar products? Visit Product Comparison Guide
General description
Hanna conductivity and TDS standards are supplied in 20 mL sachets to eliminate any chance of cross-contamination and provide the convenience of ready-to-use solutions.
Made using NIST standardized potassium chloride and silver nitrate solutions.
Made using NIST standardized potassium chloride and silver nitrate solutions.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service