Key Documents
Z618004
Binder drying oven accessories
Safety device (TWW) class 3.1
Synonym(s):
Binder 8012-0032
Select a Size
1 EA
CZK 16,000.00
CZK 16,000.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
CZK 16,000.00
About This Item
UNSPSC Code:
41104512
NACRES:
NB.24
CZK 16,000.00
Please contact Customer Service for Availability
feature
Temperature Safety Device for safe and consistent temperature regulation, Class 3.1 with visual alarm. For use with Binder units ED023 and ED400.
packaging
pack of 1 ea
availability
not available in USA
Choose from one of the most recent versions:
Certificates of Analysis (COA)