Key Documents
Z557838
Stuart melting point apparatus SMP10
AC/DC input 110 V AC
About This Item
UNSPSC Code:
41112203
NACRES:
NB.79
feature
CSA approved: no
Includes 100 melting point tubes
manufacturer/tradename
Bibby
availability
not available in EU
temperature measuring range
≤300 °C, resolution: 0.1 °C (Accuracy ± 1.0 °C at 20 °C, ± 2.5 °C at 300 °C)
General description
Designed with safety and ease of operation in mind, the SMP10 is ideal for use in education.
Two samples can be tested simultaneously and viewed via a magnifying lens aided by built in illumination.
Supplied complete with calibration certificate.
  • Digital selection and display of temperature
  • Easy and safe to operate
  • LED bulb
Legal Information
Stuart is a trademark of Bibby Sterlin Ltd.
Choose from one of the most recent versions:
Certificates of Analysis (COA)