Documents
Z531056
Sigma-Aldrich
Aldrich® Liebig condenser
Overall H 600 mm, joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint: ST/NS24/40
Overall H
600 mm
Related Categories
General description
Condensers have a standard taper female joint and a lower male drip tip-joint.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC