Key Documents
Z513105
Sigma-Aldrich
PTFE sealing washers
CGA Inlet size 170
Select a Size
1 PKG
CZK 1,540.00
CZK 1,540.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
CZK 1,540.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
CZK 1,540.00
Please contact Customer Service for Availability
packaging
pkg of 25 ea
CGA Inlet size
170
General description
Use with Aldrich lecture-bottle regulators and control valves.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Questions
1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
      Helpful?
  2. What is the product number for the PTFE washer for the Lecture Bottle Gas Regulators?
    1 answer
    1. Lecture-bottle gas regulators include a PTFE sealing washer. Spare PTFE sealing washers can be ordered.Product Z146994, PTFE sealing washers, CGA Inlet size 180 can be used for Products Z148504, Aldrich® corrosive lecture-bottle gas regulator, 0-15 psig delivery pressure, CGA Inlet size 180 and Z146714, Aldrich® non-corrosive lecture-bottle gas regulator, 0-15 psig delivery pressure, CGA Inlet size 180.Product Z513105, PTFE sealing washers, CGA Inlet size 170 can be used for Products Z512958, Aldrich® non-corrosive lecture-bottle gas regulator, 0-15 psig delivery pressure, CGA Inlet size 170 and Z513113, Aldrich® non-corrosive lecture-bottle gas regulator, 0-100 psig delivery pressure, CGA Inlet size 170.
      Helpful?