Key Documents
Z418420
Sigma-Aldrich
Aldrich® System 45 gas bubbler
About This Item
UNSPSC Code:
41121800
H
135 mm
General description
Bubbler is designed to monitor gas flow to reactions. It can be used with argon or nitrogen to provide an inert atmosphere, or gaseous reagents can be added to reactions. Supplied with (2) Viton® O-rings and (2) quick-disconnect fittings.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
System 45 is a trademark of John Babashak
Choose from one of the most recent versions:
Certificates of Analysis (COA)