Key Documents
Z414336
Sigma-Aldrich
Aldrich® septum-inlet adapter with stopcock and side-arm
Plain, joint: ST/NS 29/32, glass stopcock
Select a Size
1 EA
CZK 5,370.00
CZK 5,370.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
CZK 5,370.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
CZK 5,370.00
Please contact Customer Service for Availability
material
glass stopcock
Joint
joint: ST/NS29/32
General description
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)