Skip to Content
Merck
All Photos(1)

Key Documents

Z410144

Nalgene® vented Unitary safety wash bottle

capacity 500 mL

Synonym(s):

bottle safety, nalgene bottles, wash bottles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103209
NACRES:
NB.17

material

low-density polyethylene

feature

label Sodium hypochlorite
with cap

packaging

pkg of 4 ea

manufacturer/tradename

Nalgene® 2436-0506

capacity

500 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Unique vented closure relieves solvent vapor pressure inside wash bottle and eliminates dripping
  • Safety information printed on the bottle, including chemical color codes, chemical name and formula, hazard codes, NFPA codes, target organs, and effects and route of entry information
  • Color-coded closure
  • Easy-to-fill wide neck
  • Steady, strong dispensing stream inherent to Unitary bottle design

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Unitary is a trademark of Thermo Fisher Scientific or its subsidiaries

accessory

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service