Skip to Content
Merck
All Photos(1)

Documents

Z408522

Sigma-Aldrich

Scienceware® Cryo-Safe cold box

insulated to maintain low temperature

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123304
NACRES:
NB.22

material

polystyrene

feature

Holds 12 x 1.5 mL tubes

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F18847-0002

technique(s)

cryopreservation: suitable

L × W × H

11.7 cm (4.6 in.) × 11.7 cm (4.6 in.) × 7.1 cm (2.8 in.)

General description

Molded PS with cover. Holds twelve 1.5 mL microcentrifuge tubes. Insulated to maintain low temperature required for safe storage and transport of enzymes and critical samples. When box is taken from a 20°C freezer, it will maintain a -15°C temperature for 1.5h and will maintain a temperature below 0°C for 4h.

Legal Information

Cryo-Safe is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service