Z400564
Ace Trubore® PTFE stirrer assembly
joint: ST/NS 45/50, for flask size, 5,000 ‑ 12,000 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PTFE
Joint
joint: ST/NS45/50
feature
for flask size 5,000 ‑ 12,000 mL
packaging
pack of 1 ea
manufacturer/tradename
Ace Glass 8064-24
diam. × L
1 in. × 1 in.
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Precision PTFE Trubore stirrer assembly consisting of 10mm I.D. bearing, polished glass shaft with button, and PTFE blade. Bearing consists of PTFE inner bearing, glass-filled PTFE bushing and lock nut, compression saddle with O-ring, and threaded glass adapter.
Legal Information
Trubore is a registered trademark of Ace Glass, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service