Skip to Content
Merck
All Photos(1)

Key Documents

Z319287

Büchi® melting point apparatus

Model B-545, AC/DC input 230 V AC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112203

feature

CE compliant
Memory for 10 methods, user programmable

manufacturer/tradename

Büchi 036408

W × D × H

170 mm × 340 mm × 210 mm

Looking for similar products? Visit Product Comparison Guide

General description

For all types of melting point and boiling point determinations. Units have digital display with simple pushbutton controls, oil-free metal heating blocks, and compact footprints.
  • Accuracy: up to 100 °C (±0.3 °C); up to 250 °C (±0.5 °C); up to 400 °C (±0.8 °C)
  • 9 heating rates: 0.1, 0.2, 0.5, 1.0, 2.0, 3.0, 5.0, 10.0 and 20.0 °C/min
  • Heating: 50 to 350 °C in 7 min; Cooling: 350 to 50 °C in 10 min
  • RS-232 interface for data logging according to GLP
  • Calibration with WHO reference material possible
  • In accordance with DAB10, Ph.Eur. II, Ph.Helv.II regulations
For automatic and visual determination of three melting points and visual determination of two boiling points.

Legal Information

Büchi is a registered trademark of Büchi Labortechnik AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service