Key Documents
Z301086
Quickfit® expansion adapter
top female joint: ST/NS 14/23, bottom male joint: ST/NS 10/19
Synonym(s):
XA10
Select a Size
1 EA
CZK 1,390.00
CZK 1,390.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
CZK 1,390.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
CZK 1,390.00
Please contact Customer Service for Availability
Joint
female joint: ST/NS top14/23
male joint: ST/NS bottom10/19
manufacturer/tradename
Quickfit XA 10
availability
not available in USA and Canada
not available in USA and Canada
General description
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)