Z286419
FEP sample tube liner
Wilmad 6010-7, O.D. 10 mm
Synonym(s):
Wilmad nmr tubes, nmr sample tubes, nmr tubes
All Photos(1)
About This Item
Recommended Products
description
Recommended outer tube 513-7PP-7
packaging
pack of 1 ea
manufacturer/tradename
Wilmad 6010-7
O.D.
10 mm
volume
440 μL
Related Categories
General description
Thin-wall FEP liner minimizes filling factor losses of using inserts and protects NMR tube from etching effects of strong acids containing fluoride ions. Liners are 8 in. L and fit snugly into most common 7 in. L thin-wall tubes. A PTFE plug to close each liner is included. Not recommended for elevated temperature studies.