Skip to Content
Merck
All Photos(1)

Key Documents

Z283312

Sigma-Aldrich

Aldrich® fritted filter funnel for solid-phase synthesis

capacity 30 mL

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 11,800.00

CZK 11,800.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
CZK 11,800.00

About This Item

UNSPSC Code:
41104900
NACRES:
NB.24

CZK 11,800.00


Please contact Customer Service for Availability

material

fritted

manufacturer/tradename

Aldrich

capacity

30 mL

pore size

10—20 μm

Looking for similar products? Visit Product Comparison Guide

General description

With glass frit, 10 to 20 μm porosity, 24/40 top joint, 14/20 side joint, and gas/vacuum inlet with PTFE needle-valve and hose barb.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service