Key Documents
Z247219
Coors porcelain crucible cover
cap size B, I.D. 39 mm
Select a Size
1 PKG
CZK 5,600.00
CZK 5,600.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
CZK 5,600.00
About This Item
UNSPSC Code:
41122200
NACRES:
NB.43
CZK 5,600.00
Please contact Customer Service for Availability
cap size
B
packaging
pkg of 12 ea
manufacturer/tradename
Coorstek 66123
I.D.
39 mm
Related Categories
General description
Coors porcelain crucible cover is glazed all over except for the rim.
Features and Benefits
  • Ensures great resistance to thermal-shock
  • Durable
  • Rigid
  • Sustained refractory qualities
Other Notes
Covers may not fit exactly on crucibles due to unavoidable wall thickness variations within the crucible.
Legal Information
Coors is a trademark of Coorstek, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)