Skip to Content
Merck
All Photos(1)

Key Documents

Z242640

Whatman® disposable filter funnels

Disposable filter funnel 250 mL, 47 mm, Grade 41, 5/pk

Synonym(s):

disposable filter funnels, filter funnels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900

material

polypropylene (medical grade)

packaging

pack of 5

manufacturer/tradename

Whatman 1920-1441
Whatman Article No. 28418610 (US reference)

capacity

250 mL , 250mL reservoir

Looking for similar products? Visit Product Comparison Guide

General description

The Whatman Disposable Filter Funnels, available in 25 and 47 mm diameters, are convenient filter funnels containing Whatman brand filter media. The filter media can be easily removed for further analysis. The unit is composed of medical grade polypropylene, compatible with most solutions.

25 mm Disposable Filter Funnel

The 25 mm Disposable Filter Funnel is a convenient, disposable filter funnel containing Whatman brand filter media. This 25 mm diameter filter can be used in evaluation of processed proteins in TCA precipitation or binding assay procedures and can be easily removed for further analysis or culturing.

The 25 mm Disposable Filter Funnel is available with glass microfiber filters. Typical applications include TCA precipitation, cell harvesting, tissue washing, protein precipitation, and high recovery capture filtration.

47 mm Disposable Filter Funnel

The 47 mm Disposable Filter Funnel is a convenient, disposable unit containing Whatman brand media. The 47 mm diameter filter can be easily removed for further analysis or culturing.

Features and benefits:
  • 47 mm diameter Whatman brand filter
  • Retrievable filter for further analysis
  • Disposable for cleanliness and convenience
  • 250 mL reservoir
  • 0.45 μm cellulose nitrate available sterile for culturing

Chemical Resistance
Both 25 and 47 mm diameter filter funnels are compatible with aqueous solutions and most organic solvents. Caution should be used when working with strong acids or strong bases in these filter funnels.

Features and Benefits

Features and benefits:
  • Disposable design eliminates dedicated glassware, ideal for radioactive applications
  • Chemically resistant polypropylene housing allows for use with a wide range of aggressive solutions
  • Designed for single use or batch sample processing
  • Robotic friendly
  • Removable filter allows for further processing
  • Luer taper outlet for easy vacuum attachment

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service