Documents
Z220337
Sigma-Aldrich
Aldrich® Schlenk-type female joint cap
female joint: ST/NS 29/32
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
label
J
K
Joint
female joint: ST/NS29/32
General description
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC