Z189995
ACE heavy-wall powder funnels
Mini-Lab, angled, top diam. 60 mm
All Photos(3)
About This Item
Recommended Products
Joint
joint ST/NS14/20
manufacturer/tradename
Ace Glass 948920
top diam.
60 mm
Related Categories
General description
For pouring powders or liquids into ground-joint containers.