Key Documents
Z180351
Supelco
Tubing connector for large columns
Tubing insertion tool, tube I.D. 1.5 mm
About This Item
UNSPSC Code:
12000000
NACRES:
SB.52
manufacturer/tradename
Ace Glass 585580
technique(s)
LPLC: suitable
tube I.D.
1.5 mm
General description
TFE nut with 1/4-28 UNF 2B male thread connects small I.D. tubing to bottom drip adapter. KEL-F insert mechanically wedges tubing against fitting. For vacuum or pressure applications. Tubing-insertion tool required for assembly. Order nut, insert, and insertion tool separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)