Key Documents
Z158372
Jacketed still body, with Vigreux indents
fits Ace jacketed short path distillation apparatus
Select a Size
1 EA
CZK 19,500.00
CZK 19,500.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
CZK 19,500.00
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
CZK 19,500.00
Please contact Customer Service for Availability
manufacturer/tradename
Ace Glass 931752
General description
Replacement part for Ace jacketed short-path distillation apparatus. Vigreux indents run the entire length of the column and act as the packing for the column.
Choose from one of the most recent versions:
Certificates of Analysis (COA)