Key Documents
Z104876
PTFE sleeve
joint: ST/NS 14/35
Synonym(s):
ptfe sleeves
About This Item
UNSPSC Code:
41100000
Joint
joint: ST/NS14/35
packaging
pkg of 6 ea
manufacturer/tradename
Bel-Art
General description
Extremely thin, accurately tapered, slip-on tubes for ground-glass joints prevent seizing and galling. Chemically inert and used without lubricants or adhesives, thus precluding contamination. Temperature range: −100 to 250°C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)