WHA68752502
Whatman® GD/X syringe filters
PTFE membrane, pore size 0.2 μm, diam. 25 mm, non-sterile, pk of 1500 ea
Synonym(s):
Whatman filter, Z745332, syringe filter, syringe filters
Select a Size
CZK 97,200.00
Select a Size
About This Item
CZK 97,200.00
Recommended Products
material
PTFE membrane
polypropylene housing
sterility
non-sterile
feature
glass fiber prefilter
packaging
pk of 1500 ea
manufacturer/tradename
Whatman 6875-2502
Whatman Article No. 28420513 (US reference)
diam.
25 mm
volume
≤25 mL
pore size
0.2 μm
fitting
female Luer inlet (lock)
male Luer outlet
Related Categories
General description
Features and Benefits
- Non sterile Whatman GD/X Syringe Filters combine a polypropylene housing, high-efficiency Whatman GMF 150 and GF/F pre-filter media, and one of the following membrane materials:
- Polyethersulfone (PES) for serum, plasma and tissue culture
- Polyvinylidene difluoride (PVDF) for HPLC
- Cellulose acetate for biological solutions
- Borosilicate glass microfiber
Suitability
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)