WHA10312753
Whatman® prepleated qualitative filter paper, Grade 604 1/2
circles, diam. 320 mm, pack of 100
Synonym(s):
Z749281
Select a Size
CZK 4,470.00
Select a Size
About This Item
CZK 4,470.00
Recommended Products
material
cellulose filters
circles
packaging
pack of 100
manufacturer/tradename
Whatman 10312753
Whatman Article No. 28414304 (US reference)
parameter
50 sec/100 mL speed (Herzberg)
diam.
320 mm
pore size
25 μm
basis weight
80 g/m2
Related Categories
General description
Cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters give improved flow rate and increased loading capacity compared to equivalent quadrant folded filters.
Features and Benefits
- Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses.
- Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed.
- Increased total loading capacity as more filter area is available.
- Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself.
- The prepleating does not significantly affect any of the technical data, and the same figures may be used for the flat circles.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)