All Photos(1)
About This Item
Linear Formula:
HOC6H4CO2H
CAS Number:
Molecular Weight:
138.12
Beilstein:
970950
EC Number:
Council of Europe no.:
0693
MDL number:
UNSPSC Code:
12164502
Flavis number:
8.04
Recommended Products
Assay
≥99%
mp
213-217 °C (lit.)
SMILES string
OC(=O)c1ccc(O)cc1
InChI
1S/C7H6O3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H,(H,9,10)
InChI key
FJKROLUGYXJWQN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
B A Wols et al.
Water research, 75, 11-24 (2015-03-10)
UV/H2O2 treatment is a well-established technique to degrade organic micropollutants. A CFD model in combination with an advanced kinetic model is presented to predict the degradation of organic micropollutants in UV (LP)/H2O2 reactors, accounting for the hydraulics, fluence rate, complex
Suresh Awale et al.
Journal of natural products, 72(9), 1631-1636 (2009-08-20)
The 70% ethanol extract of Soymida febrifuga was found to kill PANC-1 human pancreatic cancer cells preferentially under nutrition-deprived conditions at a concentration of 10 microg/mL. Phytochemical investigation led to the isolation of 27 compounds including four new compounds [(3R)-6,4'-dihydroxy-8-methoxyhomoisoflavan
Céline Rivière et al.
Bioorganic & medicinal chemistry letters, 18(2), 828-831 (2007-11-29)
New polyphenol classes have been tested against amyloid-beta peptide aggregation. We have identified four novel polyphenols which could be efficient fibril inhibitors in Alzheimer's disease: malvidin and its glucoside and curculigosides B and D. We suggest that molecules with the
San-Yong Wang et al.
Bioorganic & medicinal chemistry letters, 17(10), 2785-2788 (2007-03-28)
Twelve allenic aromatic ethers, some of them are natural products isolated from the mangrove fungus Xylaria sp. 2508 in the South China Sea, were synthesized. Their antitumor activities against KB and KBv200 cells were determined. All these compounds demonstrated cytotoxic
Seth Dixon et al.
Journal of medicinal chemistry, 49(25), 7413-7426 (2006-12-08)
4-Amino-4-deoxychorismate synthase (ADCS) catalyzes the first step in the conversion of chorismate into p-aminobenzoate, which is incorporated into folic acid. We aim to discover compounds that inhibit ADCS and serve as leads for a new class of antimicrobial compounds. This
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service