W507105
Decahydro-2-naphthol
mixture of isomers, 97%
Synonym(s):
2-Decalol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C10H17OH
CAS Number:
Molecular Weight:
154.25
Beilstein:
773927
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens
biological source
synthetic
Quality Level
vapor density
>1 (vs air)
Assay
97%
refractive index
n20/D 1.5 (lit.)
bp
109 °C/14 mmHg (lit.)
density
0.996 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
Organoleptic
floral
SMILES string
OC1CCC2CCCCC2C1
InChI
1S/C10H18O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h8-11H,1-7H2
InChI key
UPMAOXLCTXPPAG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Decahydro-2-naphthol is mainly used in fragrance products.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Safety testing of fragrances: problems and implications.
Opdyke DL.
Clinical Toxicology, 10(1), 61-77 (1977)
Essential Oils and Related Products.
Guenther E, et al.
Analytical Chemistry, 37(5), 46-66 (1965)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service