Skip to Content
Merck
All Photos(1)

Key Documents

W391915

Sigma-Aldrich

2-Ethyl-6-methylpyrazine, mixture of isomers

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2
CAS Number:
Molecular Weight:
122.17
FEMA Number:
3919
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.114

biological source

synthetic

grade

Kosher

Assay

≥95%

refractive index

n20/D 1.498

density

0.968 g/mL at 25 °C

application(s)

flavors and fragrances

SMILES string

CCc1cncc(C)n1

InChI

1S/C7H10N2/c1-3-7-5-8-4-6(2)9-7/h4-5H,3H2,1-2H3

InChI key

RAFHQTNQEZECFL-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.0 °F - closed cup

Flash Point(C)

58.9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhuolin Wang et al.
Journal of agricultural and food chemistry, 68(1), 267-278 (2019-12-14)
Gas chromatography-olfactometry coupled with sensory analysis and partial least-squares regression (PLSR) analysis led to the identification of the odorants responsible for the different flavors of four yeast extracts. Sensory analysis showed that LA00L had an intense sulfurous attribute, and LA00
Julio C Page et al.
Waste management (New York, N.Y.), 69, 463-469 (2017-09-04)
Espresso capsule consumption and spent coffee ground (SCG) generation have increased, and the present study was undertaken to evaluate the volatile profile (VP), the antioxidant activity (AA) and the sun protection factor (SPF) of the Crude ethanolic extract obtained from
Yijun Liu et al.
Journal of the science of food and agriculture, 101(2), 666-672 (2020-07-23)
The volatile organic compounds (VOCs) produced from avocados during storage may be distinct at different periods, and this difference may be related to their degree of maturity, for which no relevant research has been conducted yet. A total of 30
Aileen Pua et al.
Food chemistry, 302, 125370-125370 (2019-08-24)
Four Arabica coffees (Brazil, Colombia, Ethiopia, and Guatemala) yield highly variant odours, attesting to the complexities of coffee aroma that command advanced analytical tools. In this study, their volatiles were extracted using solvent-assisted flavour evaporation (SAFE) and headspace solid-phase microextraction
Jie Liu et al.
Food chemistry, 345, 128826-128826 (2021-02-20)
Brown sugar (non-centrifugal cane sugar) is popular for its pleasant caramel-like aroma and sweetness. Vacuum simultaneous steam distillation and extraction (V-SDE) and gas chromatography-mass spectrometry (GC-MS) was used to study the volatile fraction of brown sugar. To further determine the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service