Skip to Content
Merck
All Photos(1)

Key Documents

W341901

Sigma-Aldrich

1,2-Propanediol dibenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO2CH2CH(CH3)O2CC6H5
CAS Number:
Molecular Weight:
284.31
FEMA Number:
3419
EC Number:
MDL number:
UNSPSC Code:
12164502
Flavis number:
9.803

refractive index

n20/D 1.545 (lit.)

bp

232 °C/12 mmHg (lit.)

mp

−3 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

SMILES string

CC(COC(=O)c1ccccc1)OC(=O)c2ccccc2

InChI

1S/C17H16O4/c1-13(21-17(19)15-10-6-3-7-11-15)12-20-16(18)14-8-4-2-5-9-14/h2-11,13H,12H2,1H3

InChI key

UMVMVEZHMZTUHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service