Skip to Content
Merck
All Photos(1)

Key Documents

W326100

Sigma-Aldrich

2-sec-Butylcyclohexanone

mixture of diastereomers, ≥98%, FG

Synonym(s):

peppermint cyclohexanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)C6H9(=O)
CAS Number:
Molecular Weight:
154.25
FEMA Number:
3261
Beilstein:
1859137
EC Number:
Council of Europe no.:
11044
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.095
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥98%

refractive index

n20/D 1.458 (lit.)

bp

76-78 °C/8 mmHg (lit.)

density

0.912 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cooling; woody; camphoraceous; minty; peppermint

SMILES string

CCC(C)C1CCCCC1=O

InChI

1S/C10H18O/c1-3-8(2)9-6-4-5-7-10(9)11/h8-9H,3-7H2,1-2H3

InChI key

RQXTZKGDMNIWJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-sec-Butylcyclohexanone can be used as a flavoring agent[1] and fragrance ingredient.[2]

Biochem/physiol Actions

Taste at 20 ppm

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flavoring Ingredients Classified As GRAS By the Flavor Extract Manufacturers Association.
Source Book of Flavors, 655-670 (1995)
A toxicologic and dermatologic assessment of cyclic acetates when used as fragrance ingredients
Belsito D, et al.
Food And Chemical Toxicology, 46(12), S1-S27 (2008)
Mosciano, G.
Perfumer & Flavorist, 30, 53-53 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service