Skip to Content
Merck
All Photos(1)

Documents

W262501

Sigma-Aldrich

Lemon oil

rectified, Argentina origin, FG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2625
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Citrus limon (L.) Burm. F.

grade

FG
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.20

optical activity

[α]20/D +75°, neat

origin

Argentina origin

refractive index

n20/D 1.473 (lit.)

bp

176 °C (lit.)

density

0.850 g/mL at 25 °C
0.852 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

lemon; citrus

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Extraction method: cold pressed

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service