Skip to Content
Merck
All Photos(1)

Key Documents

THS03131

ThalesNano CatCart® catalyst cartridge system, 70 mm L

NP Pd(0) EnCat® 30

Synonym(s):

Heterogeneous catalysis system, safe Heterogeneous catalysis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103817
NACRES:
NB.51
Pricing and availability is not currently available.

packaging

pkg of 1 ea

manufacturer/tradename

ThalesNano THS 03131

diam.

4 mm

Looking for similar products? Visit Product Comparison Guide

General description

CatCarts are suitable for use only with Thalesnano H-Cube reactors.

Legal Information

CatCart is a registered trademark of ThalesNano Nanotechnology Inc.
EnCat is a registered trademark of Reaxa Ltd.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Sol. 1 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation

Target Organs

Nervous system

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service