Key Documents
TEM-FF400CUUL
Sigma-Aldrich
Ultrathin Formvar Supported Copper Square Mesh
size 400 mesh, box of 50
Synonym(s):
FORMVAR SQUARE MESH - CU 400 MESH UL 50/BX
About This Item
UNSPSC Code:
41111761
packaging
box of 50
size
400 mesh
General description
A 5-6 nm ultrathin formvar film supported standard copper square mesh grids. Copper grid, 400 mesh. All of our film is laid on the shiny side of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)