Documents
TEM-CFT400CU
Sigma-Aldrich
Carbon Film Supported Copper Thin Bar Square Mesh Grid
size 400 mesh, box of 25
Synonym(s):
CARBON FILM ONLY ON THIN BAR 400 MESH COPPER GRIDS 25/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 25
size
400 mesh
General description
A 5-6 nm thin film of pure carbon supported thin bar square mesh grid. Copper grid, 400 mesh. Carbon film show better electrical and heat conduction in the electron beam.