Key Documents
TEM-CFLF135-AU-25
Sigma-Aldrich
Carbon Film Supported Gold London Finder LF135 Grids
box of 25
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-CFLF135-AU-25 is not currently sold in your country. Contact Technical Service
packaging
box of 25
General description
A 5-6 nm thin film of pure carbon supported London finder (LF135) grids. Gold grid. Carbon film show better electrical and heat conduction in the electron beam. London finder grids are locator grids with reference/index on the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)