Key Documents
TEM-3750
Sigma-Aldrich
CryoCapsule
3 mm, pack of 10
About This Item
UNSPSC Code:
41111761
packaging
pack of 10
General description
The CryoCapsule is a new tool that is compatible with High Pressure Freezing (HPF). The CryoCapsule is dedicated to correlative light to electron microscopy experiments. Comparable to a small petri dish, it is composed of a landmarked sapphire disc and
Choose from one of the most recent versions:
Certificates of Analysis (COA)