Skip to Content
Merck
All Photos(1)

Key Documents

T14400

Sigma-Aldrich

Tetrahydropyran

ReagentPlus®, 99%

Synonym(s):

Pentamethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O
CAS Number:
Molecular Weight:
86.13
Beilstein:
102436
EC Number:
MDL number:
UNSPSC Code:
12190000

vapor density

3 (vs air)

product line

ReagentPlus®

Assay

99%

refractive index

n20/D 1.42 (lit.)

bp

88 °C (lit.)

mp

−45 °C (lit.)

density

0.881 g/mL at 25 °C (lit.)

SMILES string

C1CCOCC1

InChI

1S/C5H10O/c1-2-4-6-5-3-1/h1-5H2

InChI key

DHXVGJBLRPWPCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

3.2 °F - closed cup

Flash Point(C)

-16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B V Subba Reddy et al.
Organic & biomolecular chemistry, 13(24), 6737-6741 (2015-05-23)
A domino reaction has been developed for the synthesis of oxygen bridged bicyclic ethers through the coupling of 4-(2-hydroxyethyl)cyclohex-3-enols with aldehydes in the presence of 10 mol% of molecular iodine in dichloromethane at 25 °C. This method is highly diastereoselective
Yantao Wei et al.
Retina (Philadelphia, Pa.), 35(8), 1631-1639 (2015-07-28)
To evaluate the surgical approach of pars plana vitrectomy combined with 360° retinotomy and silicon oil tamponade in the treatment of patients with large subretinal hemorrhage. Prospective, nonrandomized, and noncomparative case series study. Consecutive patients with breakthrough vitreous hemorrhage and
Paul A Clarke et al.
Organic & biomolecular chemistry, 13(16), 4743-4750 (2015-03-26)
The Maitland-Japp reaction has been extended to the synthesis of highly functionalised dihydropyran-4-ones. These dihydropyran-4-ones can in turn be converted stereoselectively into tetrahydropyran-4-ones with tertiary and quaternary stereocentres via the one-pot addition of hydride or carbon nucleophiles and trapping with
Francesco Paolo Di Nicola et al.
Dalton transactions (Cambridge, England : 2003), 44(28), 12653-12659 (2015-06-17)
Distinct batches of orange (1a-e) and green crystals (2a-e) were isolated from the reactions of MoCl5 with tetrahydropyran (thp), respectively at room temperature (in CH2Cl2) and at ca. 80 °C (in ClCH2CH2Cl). Crystals 2a-e are isomorphous to 1a-e and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service