Merck
Skip To
SYNC174200
Synthware water jacketed distillation column with indents to support packing material
joint: ST/NS 24/40, jacket H 200 mm
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
2 640,00 CZK
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
Manufacturer/tradename:
Synthware C174200
Packaging:
pkg of 1 ea
Joint:
joint: ST/NS24/40
2 640,00 CZK
Please contact Customer Service for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C174200
jacket H
200 mm
General description
Indentations at the bottom for supporting packing material. Jacket permits the column to be used as a condenser.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
SYNC171120SYNC171200SYNC179110
manufacturer/tradename
Synthware C174200
manufacturer/tradename
Synthware C171120
manufacturer/tradename
Synthware C171200
manufacturer/tradename
Synthware C179110
joint
joint: ST/NS24/40
joint
joint: ST/NS14/20
joint
joint: ST/NS14/20
joint
joint: ST/NS19/22
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
jacket H
200 mm
jacket H
110 mm
jacket H
200 mm
jacket H
110 mm
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)