Documents
SYNA262419
Synthware Kjeldahl distilling trap
100 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 19/22
Synonym(s):
Synthware A262419, Distilling trap, Kjeldahl trap
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
bottom joint: ST/NS19/22
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A262419
capacity
100 mL
Related Categories
General description
Adapter is placed above flask to prevent carry-over of reactions which have a tendency to foam.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Visit the Document Library
Articles
Learn how water impurities can impact the Kjeldahl nitrogen method. Get recommendations for accurate Kjeldahl analysis and total Kjeldahl nitrogen (TKN) tests. Select the optimal water purification system.