SLW1450/06D
Pyrex® Evaporating dish
170 mL
Synonym(s):
Pyrex, SciLabware 1450/06D
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
flat base
packaging
pack of 10 ea
manufacturer/tradename
SciLabware 1450/06D
H
55 mm
diam.
95 mm
dish diam.
95 mm
capacity
170 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Complies to DIN 12336
- Manufactured from Pyrex borosilicate glass
- High resistance to chemical attack and thermal shock
- Flat base
- Curved sides with spout
Features and Benefits
• Complies to DIN 12336• Manufactured from borosilicate glass 3.3• Curved sides with spout• Flat base• High resistance to chemical attack and thermal shock• 1450/01D, 1450/03D & 1450/04D are unbadged.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service