Skip to Content
Merck
All Photos(1)

Documents

QBD10201

Sigma-Aldrich

Biotin-dPEG®3-MAL

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H43N5O8S
Molecular Weight:
597.72
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reaction type: Biotinylations
reactivity: thiol reactive

reagent type: linker

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(NCCCOCCOCCOCCCNC(CCCCC1C2NC(NC2CS1)=O)=O)CCN3C(C=CC3=O)=O

Application

Biotin-dPEG®3-MAL can be used as a cross-linking reagent in biotinylation reaction.

Features and Benefits

Biotin-dPEG®3-MAL takes advantage of the well-known, specific Michael reaction between a maleimide group and a free sulfhydryl to allow site-specific biotin labeling of molecules in aqueous media within the pH range of 6.5 – 7.5. The single molecular weight dPEG® spacer (24.9 Å) provide precise spacing between the biotin moiety and the labeled molecule. The dPEG® spacer imparts water solubility to the labeled molecule while reducing immunogenicity, aggregation, and precipitation of labeled biomolecules or other labeled compounds.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda R Smith et al.
Retrovirology, 16(1), 28-28 (2019-10-24)
PIE12-trimer is a highly potent D-peptide HIV-1 entry inhibitor that broadly targets group M isolates. It specifically binds the three identical conserved hydrophobic pockets at the base of the gp41 N-trimer with sub-femtomolar affinity. This extremely high affinity for the transiently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service