Skip to Content
Merck
All Photos(1)

Key Documents

P53306

Sigma-Aldrich

Propyl gallate

97%

Synonym(s):

3,4,5-Trihydroxybenzoic acid propyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3,4,5-(HO)3C6H2CO2CH2CH2CH3
CAS Number:
Molecular Weight:
212.20
Beilstein:
1877976
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

97%

mp

146-149 °C (lit.)

SMILES string

CCCOC(=O)c1cc(O)c(O)c(O)c1

InChI

1S/C10H12O5/c1-2-3-15-10(14)6-4-7(11)9(13)8(12)5-6/h4-5,11-13H,2-3H2,1H3

InChI key

ZTHYODDOHIVTJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Reconstitution

Prepare a 0.1M solution in glycerol:PBS (9:1)

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

368.6 °F - closed cup

Flash Point(C)

187 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tanvir Hussain et al.
European journal of medicinal chemistry, 44(11), 4654-4660 (2009-08-12)
A series of new 3-[4-(1H-imidazol-1-yl) phenyl]prop-2-en-1-ones were synthesized by the condensation of various acetophenones with 4-(1H-imidazol-1-yl) benzaldehyde which was itself prepared by the N-arylation of imidazole using hexadecyltrimethylammonium bromide as catalyst for the first time. All the synthesized compounds were
Matloob Ahmad et al.
European journal of medicinal chemistry, 45(2), 698-704 (2009-12-08)
A series of potential anti-oxidant and anti-bacterial N'-arylmethylidene-2-(3,4-dimethyl-5,5-dioxidopyrazolo[4,3-c][1,2]benzothiazin-2(4H)-yl)acetohydrazides was synthesized in a facile way starting from commercially available saccharine. 3,4-Dimethyl-2,4-dihydropyrazolo[4,3-c][1,2]benzothiazine 5,5-dioxide was obtained by ring expansion of 2-(2-oxopropyl)-1,2-benzisothiazol-3(2H)-one 1,1-dioxide followed by N-methylation and cyclization with hydrazine using ultrasonic irradiation. N-alkylation
Imtiaz Khan et al.
European journal of medicinal chemistry, 45(11), 5200-5207 (2010-09-11)
New series of 4,5-disubstituted-2,4-dihydro-3H-1,2,4-triazole-3-thiones (8a-j) and 2,5-disubstituted-1,3,4-thiadiazoles (9a-h) were synthesized by dehydrative cyclization of hydrazinecarbothioamide derivatives (7a-k) by refluxing in 4N aqueous sodium hydroxide and by overnight stirring with polyphosphoric acid, respectively. The structures of the newly synthesized compounds were
Kathrin Schneider et al.
Journal of natural products, 72(10), 1768-1772 (2009-10-24)
A family of new secondary metabolites with a carbazole moiety and an alkyl side chain was isolated from Tsukamurella pseudospumae strain Acta 1857. They were named lipocarbazoles in accordance with their chemical structures, which were determined by mass spectrometry and
P C Leal et al.
Bioorganic & medicinal chemistry letters, 19(6), 1793-1796 (2009-02-24)
The antifungal activity of a complete series of 15 n-alkyl gallates and six analogues acting against a representative panel of opportunistic pathogenic fungi was studied in order to analyze their role in: the importance of the fungi tested, the importance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service