Skip to Content
Merck
All Photos(1)

Key Documents

P4003

Sigma-Aldrich

Pentadecylamine

96%

Synonym(s):

1-Aminopentadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14NH2
CAS Number:
Molecular Weight:
227.43
Beilstein:
1744557
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
96%

vapor pressure

1 mmHg ( 120.5 °C)

Assay

96%

form

solid

bp

299-301 °C (lit.)

mp

35-37 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCN

InChI

1S/C15H33N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16/h2-16H2,1H3

InChI key

JPZYXGPCHFZBHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mireille Alhouayek et al.
Scientific reports, 10(1), 6314-6314 (2020-04-15)
There is good evidence that the N-acylethanolamine (NAE)/monoacylglycerol (MAG) signalling systems are involved in the pathogenesis of cancer. However, it is not known how prostate tumours affect these systems in the surrounding non-malignant tissue and vice versa. In the present
Mireille Alhouayek et al.
British journal of pharmacology, 176(10), 1470-1480 (2018-01-10)
Endocannabinoids and related N-acylethanolamines (NAEs) are involved in regulation of gut function, but relatively little is known as to whether inflammatory cytokines such as IFNγ affect their levels. We have investigated this in vitro using cultures of T84 colon cancer
Sanaz Hashemian et al.
Innate immunity, 23(8), 687-696 (2017-09-30)
TLR4 respond to bacterial LPS to produce inflammatory cytokines. TLR4 are expressed in dorsal root ganglia and play a role in pain. F11 dorsal root ganglia × mouse neuroblastoma cells possess many of the properties seen in nociceptive dorsal root

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service