Skip to Content
Merck
All Photos(1)

Documents

N33101

Sigma-Aldrich

Norharmane

98%

Synonym(s):

β-Carboline, 9H-Pyrido[3,4-b]indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8N2
CAS Number:
Molecular Weight:
168.19
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

98%

SMILES string

c1ccc2c(c1)[nH]c3cnccc23

InChI

1S/C11H8N2/c1-2-4-10-8(3-1)9-5-6-12-7-11(9)13-10/h1-7,13H

InChI key

AIFRHYZBTHREPW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitor of indoleamine 2,3-dioxygenase (IDO).

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Micaela Gonzalez et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 8(8), 1139-1149 (2009-07-30)
The photochemistry of norharmane (9H-pyrido[3,4-b]indole) in acidic (pH 5.0+/-0.1) and alkaline (pH 10.0+/-0.1) aqueous solutions was studied. The photochemical reactions were monitored by TLC, UV/VIS absorption spectroscopy, high-performance liquid chromatography (HPLC), electronic ionization-mass spectrometry (EI-MS), UV-laser desorption/ionization-time of flight-mass spectrometry
Rosa Busquets et al.
Molecular nutrition & food research, 53(12), 1496-1504 (2009-10-08)
Heterocyclic amines (HCAs) are potent mutagens/carcinogens to which humans are frequently exposed through the consumption of cooked meat and fish food. The effect of normal intake of HCAs and their role in the aetiology of human cancer is unknown. To
Olga I Tarzi et al.
Journal of mass spectrometry : JMS, 44(2), 260-277 (2008-11-18)
The thermal stability of several commonly used crystalline matrix-assisted ultraviolet laser desorption/ionization mass spectrometry (UV-MALDI-MS) matrices, 2,5-dihydroxybenzoic acid (gentisic acid; GA), 2,4,6-trihydroxyacetophenone (THA), alpha-cyano-4-hydroxycinnamic acid (CHC), 3,5-dimethoxy-4-hydroxycinnamic acid (sinapinic acid; SA), 9H-pirido[3,4-b]indole (nor-harmane; nor-Ho), 1-methyl-9H-pirido[3,4-b]indole (harmane; Ho), perchlorate of nor-harmanonium
Andrés G León et al.
Journal of chromatography. A, 1192(2), 254-258 (2008-04-25)
The presence of cyclodextrins (CDs) in the mobile phase alters the chromatographic equilibria and induces a secondary chemical equilibrium associated to the chromatographic separation by HPLC. In this study the influence of the presence of CDs in the mobile phase
Ouafa Arib et al.
European journal of pharmacology, 629(1-3), 47-52 (2009-12-23)
It has been suggested that the beta-carbolines harmane and norharmane may be involved in the pathophysiology of Parkinson's disease, psychosis and addiction, but the mechanisms of these possible effects remain to be elucidated. In the present study, the effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service