All Photos(1)
About This Item
Linear Formula:
O2NC6H4CH2COCO2H
CAS Number:
Molecular Weight:
209.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
powder
mp
119-120 °C (lit.)
SMILES string
OC(=O)C(=O)Cc1ccccc1[N+]([O-])=O
InChI
1S/C9H7NO5/c11-8(9(12)13)5-6-3-1-2-4-7(6)10(14)15/h1-4H,5H2,(H,12,13)
InChI key
CGCWRLDEYHZQCW-UHFFFAOYSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service