Skip to Content
Merck
All Photos(1)

Key Documents

N15006

Sigma-Aldrich

3-Nitrobiphenyl

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4NO2
CAS Number:
Molecular Weight:
199.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

58-60 °C (lit.)

SMILES string

[O-][N+](=O)c1cccc(c1)-c2ccccc2

InChI

1S/C12H9NO2/c14-13(15)12-8-4-7-11(9-12)10-5-2-1-3-6-10/h1-9H

InChI key

FYRPEHRWMVMHQM-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xia Wang et al.
Talanta, 186, 561-567 (2018-05-23)
Metal-organic nanotubes (MONTs) have great potential for solid phase microextraction (SPME) on account of high specific surface area, tunable pore sizes, and exceptional thermal and chemical stability. In this work, zinc (II)-based metal-organic nanotubes (Zn-MONTs) were used as an adsorbent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service