All Photos(1)
About This Item
Linear Formula:
(CH3)2CHCH2CH=CH2
CAS Number:
Molecular Weight:
84.16
Beilstein:
1731096
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
>1 (vs air)
vapor pressure
4.45 psi ( 20 °C)
Assay
98%
autoignition temp.
572 °F
refractive index
n20/D 1.382 (lit.)
bp
53-54 °C (lit.)
density
0.665 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CC(C)CC=C
InChI
1S/C6H12/c1-4-5-6(2)3/h4,6H,1,5H2,2-3H3
InChI key
WSSSPWUEQFSQQG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
accessory
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-23.8 °F
Flash Point(C)
-31 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Esther Marco et al.
Journal of chromatography. A, 1410, 51-59 (2015-08-06)
A method for the rapid analysis of volatile organic compounds (VOCs) in smoke from tobacco and electronic cigarettes and in exhaled breath of users of these smoking systems has been developed. Both disposable and rechargeable e-cigarettes were considered. Smoke or
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service