Skip to Content
Merck
All Photos(1)

Documents

L550001-HH

Sigma-Aldrich

Cap B2

configured for ÄKTA® and OligoPilot®

Synonym(s):

Cap B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

type

for DNA synthesis

product line

Proligo Reagents

packaging

pkg of 1 × 1.25 L (L550251-01)

impurities

≤200 ppm water content (Karl Fischer)

compatibility

configured for ÄKTA® and OligoPilot®

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Acetonitrile / Pyridine 6/4 (v/v)

Legal Information

OligoPilot is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F

Flash Point(C)

17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service