H31328
2-Hydroxy-9-fluorenone
98%
Synonym(s):
NSC 22835
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C13H8O2
CAS Number:
Molecular Weight:
196.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
impurities
<20% 2-fluoro-9-fluorenone
mp
204-206 °C (lit.)
SMILES string
Oc1ccc2-c3ccccc3C(=O)c2c1
InChI
1S/C13H8O2/c14-8-5-6-10-9-3-1-2-4-11(9)13(15)12(10)7-8/h1-7,14H
InChI key
GXUBPHMYNSICJC-UHFFFAOYSA-N
Application
Reactant involved in:
- One-pot synthesis of aryl substituted aurones
- Studies to tune HOMO-LUMO levels of 9-fluorenone scaffolds
- O-methylation for synthesis of second-generation light driven rotary molecular motors
- Reactions with aluminum, gallium compounds, and methylquinolines
- Synthesis of acridinyl- and fluorenylidenehydrazido calixarenes
- Acetylation for synthesis of acetates
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Rozanna Avagyan et al.
Talanta, 165, 702-708 (2017-02-06)
Up until now, the methods used for determination of hydroxylated polycyclic aromatic hydrocarbons in air particulate samples have been target methods, only determining compounds with available reference standards. In this present study, a combined target and suspect screening strategy for
Rozanna Avagyan et al.
Analytical and bioanalytical chemistry, 407(16), 4523-4534 (2015-05-04)
A simple and fast method for analysis of hydroxylated polycyclic aromatic hydrocarbons using pressurized liquid extraction and high performance liquid chromatography utilizing photoionization tandem mass spectrometry was developed. Simultaneous separation and determination of nine hydroxylated polycyclic aromatic hydrocarbons and two
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service