Skip to Content
Merck
All Photos(1)

Key Documents

GF96026418

Quartz - fused

sheet, thickness 3.0 mm, size 50 × 50 mm, condition ground

Synonym(s):

SI613142

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 11,100.00

CZK 11,100.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
CZK 11,100.00

About This Item

UNSPSC Code:
11101503
NACRES:
NA.23

CZK 11,100.00


Please contact Customer Service for Availability

Request a Bulk Order

form

sheet

manufacturer/tradename

Goodfellow 960-264-18

size

50 × 50 mm

thickness

3.0 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service