Skip to Content
Merck
All Photos(1)

Key Documents

GF77604059

Monel® alloy 400, Ni65Cu33Fe2

mesh, 300 x 300mm, 0.38mm nominal aperture, 0.25mm wire diameter, 40 x 40 wires/inch, 0.37 open area, plain weave mesh

Synonym(s):

NI118710

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23
To inquire about this product GF77604059, please contact your local Merck office or dealer. Contact Technical Service

description

40 x 40 (wires/inch)

form

(mesh)

manufacturer/tradename

Goodfellow 776-040-59

diam. × nominal capacity

0.25 mm × 0.38 mm , plain weave mesh

size × opening

300 mm × 300 mm × 0.37

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Monel is a registered trademark of Special Metals Corporation
Monel is a registered trademark of Huntington Alloys Corp.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service