Skip to Content
Merck
All Photos(2)

Key Documents

GF68735065

Hafnium

rod, 500mm, diameter 2.0mm, as drawn, 97%

Synonym(s):

Hafnium, HF007902

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 27,800.00

CZK 27,800.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
CZK 27,800.00

About This Item

Empirical Formula (Hill Notation):
Hf
CAS Number:
Molecular Weight:
178.49
MDL number:
UNSPSC Code:
12141718
PubChem Substance ID:
NACRES:
NA.23

CZK 27,800.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

rod

manufacturer/tradename

Goodfellow 687-350-65

resistivity

29.6 μΩ-cm, 0°C

L × diam.

500 mm × 2.0 mm

bp

4602 °C (lit.)

mp

2227 °C (lit.)

density

13.3 g/cm3 (lit.)

SMILES string

[Hf]

InChI

1S/Hf

InChI key

VBJZVLUMGGDVMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service